Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102171003
Entrez Gene ID
102171003
Gene Name
V-set and transmembrane domain-containing protein 1
Gene Aliases
VSTM1
Chromosome Location
Chr.18: 63432319 - 63454661 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692929.3 XP_005692986.1 2 - 3 261 86
GenBank mRNA JO319208.2 - 2 - 3 247 86

View Full Product Details