Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPX
Entrez Gene ID
102171850
Gene Name
spexin hormone
Gene Aliases
C5H12orf39
Chromosome Location
Chr.5: 87234889 - 87243809 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048410.1 XP_017903899.1 4 - 5 515 99
XM_018048411.1 XP_017903900.1 4 - 5 288 99

View Full Product Details