Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELOC
Entrez Gene ID
102182517
Gene Name
elongin C
Gene Aliases
TCEB1
Chromosome Location
Chr.14: 44881006 - 44900556 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689072.3 XP_005689129.1 3 - 4 382 106
XM_018058400.1 XP_017913889.1 3 - 4 410 106
GenBank mRNA JO611214.2 - 1 - 2 415 106

View Full Product Details