Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG3
Entrez Gene ID
102172403
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
Chromosome Location
Chr.25: 21863693 - 21962066 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697611.3 XP_005697668.1 2 - 3 1498 104

View Full Product Details