Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIPC3
Entrez Gene ID
102174831
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
-
Chromosome Location
Chr.7: 90218056 - 90221460 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050828.1 XP_017906317.1 5 - 6 827 77
GenBank mRNA JT634948.1 - 3 - 4 1803 77

View Full Product Details