Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSTN
Entrez Gene ID
102187781
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
-
Chromosome Location
Chr.13: 37403918 - 37429845 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057224.1 XP_017912713.1 3 - 4 526 109
GenBank mRNA JO643404.2 - 3 - 4 727 109

View Full Product Details