Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
102178250
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 36596053 - 36600419 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044316.1 XP_017899805.1 1 - 2 1390 66
GenBank mRNA JT662117.1 - 4 - 5 529 66

View Full Product Details