Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL1F10
Entrez Gene ID
102182135
Gene Name
interleukin 1 family member 10
Gene Aliases
-
Chromosome Location
Chr.11: 46412353 - 46423768 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686679.2 XP_005686736.2 4 - 5 311 85
XM_005686680.3 XP_005686737.2 4 - 5 356 85
XM_018054765.1 XP_017910254.1 3 - 4 267 85
XM_018054766.1 XP_017910255.1 3 - 4 241 85
GenBank mRNA KA350578.1 - 4 - 5 307 85

View Full Product Details