Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XKR9
Entrez Gene ID
102178186
Gene Name
XK related 9
Gene Aliases
-
Chromosome Location
Chr.14: 47740303 - 47760662 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689056.2 XP_005689113.1 1 - 2 280 107

View Full Product Details