Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL27
Entrez Gene ID
102172191
Gene Name
ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.19: 42699567 - 42702237 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693883.3 XP_005693940.1 3 - 4 361 122
XM_005693884.3 XP_005693941.1 3 - 4 361 122
XM_005693885.3 XP_005693942.1 3 - 4 458 122
XM_018065160.1 XP_017920649.1 3 - 4 459 122
GenBank mRNA KA320940.1 - 2 - 3 302 122

View Full Product Details