Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTRA4
Entrez Gene ID
102180823
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
Chromosome Location
Chr.27: 11582000 - 11594064 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042082.1 XP_017897571.1 2 - 3 645 63

View Full Product Details