Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
C14H8orf82
Entrez Gene ID
102174407
Gene Name
chromosome 14 C8orf82 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 81515491 - 81518595 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013969043.2 XP_013824497.1 1 - 2 796 90
GenBank mRNA JO384680.2 - 1 - 2 200 90

View Full Product Details