The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018055628.1 | XP_017911117.1 | 3 - 4 | 979 | 108 |
GenBank mRNA | JO646145.2 | - | 3 - 4 | 1044 | 108 |