Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPFFR1
Entrez Gene ID
102183209
Gene Name
neuropeptide FF receptor 1
Gene Aliases
-
Chromosome Location
Chr.28: 19175778 - 19197376 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042160.1 XP_017897649.1 3 - 4 538 65

View Full Product Details