Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPZ
Entrez Gene ID
102189639
Gene Name
myelin protein zero
Gene Aliases
-
Chromosome Location
Chr.3: 111905320 - 111910331 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677158.2 XP_005677215.2 4 - 5 639 96
GenBank mRNA KA346345.1 - 4 - 5 637 96

View Full Product Details