Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GKN2
Entrez Gene ID
102186315
Gene Name
gastrokine 2
Gene Aliases
-
Chromosome Location
Chr.11: 67025322 - 67033288 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686830.3 XP_005686887.1 4 - 5 368 94

View Full Product Details