Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf70
Entrez Gene ID
102187054
Gene Name
chromosome 11 C2orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 72709880 - 72727597 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686929.1 XP_005686986.1 1 - 2 113 70

View Full Product Details