The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_013966432.2 | XP_013821886.2 | 3 - 4 | 274 | 103 |
GenBank mRNA | JO644545.2 | - | - | 433 | 103 |