Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX15
Entrez Gene ID
102175552
Gene Name
testis expressed 15, meiosis and synapsis associated
Gene Aliases
-
Chromosome Location
Chr.27: 18654420 - 18671829 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041766.1 XP_017897255.1 4 - 5 8637 73

View Full Product Details