Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTE1
Entrez Gene ID
102176735
Gene Name
asteroid homolog 1
Gene Aliases
-
Chromosome Location
Chr.1: 138467430 - 138489608 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675616.3 XP_005675673.2 5 - 6 2405 77
XM_005675617.3 XP_005675674.2 5 - 6 2002 77
XM_005675618.3 XP_005675675.2 4 - 5 2234 77
XM_013966561.2 XP_013822015.2 5 - 6 2043 77

View Full Product Details