Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC12
Entrez Gene ID
102179017
Gene Name
zinc finger DHHC-type containing 12
Gene Aliases
-
Chromosome Location
Chr.11: 98756396 - 98759892 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687208.3 XP_005687265.1 1 - 2 143 104
GenBank mRNA JO584870.2 - 1 - 2 117 104
KA316541.1 - 1 - 2 135 104

View Full Product Details