Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2I
Entrez Gene ID
102172473
Gene Name
RNA polymerase II subunit I
Gene Aliases
-
Chromosome Location
Chr.18: 47615108 - 47616741 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692272.3 XP_005692329.1 5 - 6 746 78
XM_013971226.1 XP_013826680.1 4 - 5 597 78
GenBank mRNA JO445792.2 - 5 - 6 695 78

View Full Product Details