Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP68
Entrez Gene ID
102179777
Gene Name
centrosomal protein 68
Gene Aliases
-
Chromosome Location
Chr.11: 63174597 - 63201889 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686803.3 XP_005686860.1 4 - 5 3071 76
XM_018055427.1 XP_017910916.1 5 - 6 2236 76
XM_018055428.1 XP_017910917.1 5 - 6 2236 76
GenBank mRNA JO631208.2 - 5 - 6 2254 76
JO631212.2 - 5 - 6 2254 76

View Full Product Details