Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYT9
Entrez Gene ID
102171629
Gene Name
synaptotagmin 9
Gene Aliases
-
Chromosome Location
Chr.15: 36940602 - 37152007 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059486.1 XP_017914975.1 4 - 5 1384 80
XM_018059487.1 XP_017914976.1 4 - 5 1384 80

View Full Product Details