Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKAIN3
Entrez Gene ID
102190049
Gene Name
sodium/potassium transporting ATPase interacting 3
Gene Aliases
-
Chromosome Location
Chr.14: 54129545 - 54655603 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058468.1 XP_017913957.1 4 - 5 3545 113
GenBank mRNA JO617467.2 - 2 - 3 114 113
JO617468.2 - 2 - 3 114 113
JT721206.1 - 1 - 2 35 113

View Full Product Details