Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO43
Entrez Gene ID
102190619
Gene Name
F-box protein 43
Gene Aliases
-
Chromosome Location
Chr.14: 17958637 - 17974191 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058083.1 XP_017913572.1 5 - 6 2187 114

View Full Product Details