Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C13H20orf85
Entrez Gene ID
102191811
Gene Name
chromosome 13 C20orf85 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 57478806 - 57494461 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688312.1 XP_005688369.1 2 - 3 206 94

View Full Product Details