Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD1
Entrez Gene ID
102189024
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 97001714 - 97004975 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051194.1 XP_017906683.1 2 - 3 1584 102
GenBank mRNA JO332714.2 - 3 - 4 922 102

View Full Product Details