Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRF1
Entrez Gene ID
102171130
Gene Name
perforin 1
Gene Aliases
-
Chromosome Location
Chr.28: 18870678 - 18875354 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699151.3 XP_005699208.1 2 - 3 673 119
GenBank mRNA JO331984.2 - 1 - 2 1765 119
JO595767.2 - 2 - 3 657 119

View Full Product Details