Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO33
Entrez Gene ID
102173044
Gene Name
F-box protein 33
Gene Aliases
-
Chromosome Location
Chr.21: 48697385 - 48717234 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973125.2 XP_013828579.1 2 - 3 1054 86

View Full Product Details