Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPTN
Entrez Gene ID
102188911
Gene Name
repetin
Gene Aliases
-
Chromosome Location
Chr.3: 101978453 - 101984509 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677580.2 XP_005677637.2 2 - 3 199 81
GenBank mRNA KA311339.1 - 1 - 2 375 81

View Full Product Details