The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018051585.1 | XP_017907074.1 | 5 - 6 | 800 | 88 |
XM_018051586.1 | XP_017907075.1 | 4 - 5 | 598 | 88 | |
GenBank mRNA | KA308604.1 | - | 4 - 5 | 704 | 88 |