Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM47
Entrez Gene ID
102175380
Gene Name
RNA binding motif protein 47
Gene Aliases
-
Chromosome Location
Chr.6: 59968310 - 60138027 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681536.3 XP_005681593.1 7 - 8 1987 88
XM_005681537.3 XP_005681594.1 5 - 6 2158 88
XM_005681538.3 XP_005681595.1 6 - 7 1854 88
XM_005681539.3 XP_005681596.1 4 - 5 1561 88
XM_013964599.2 XP_013820053.1 4 - 5 1543 88
XM_013964600.2 XP_013820054.1 4 - 5 1575 88
XM_013964601.2 XP_013820055.1 5 - 6 2156 88
XM_018049385.1 XP_017904874.1 5 - 6 1945 88
XM_018049386.1 XP_017904875.1 6 - 7 2257 88
GenBank mRNA JO429085.2 - 2 - 3 400 88

View Full Product Details