Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGRP
Entrez Gene ID
102183996
Gene Name
agouti related neuropeptide
Gene Aliases
-
Chromosome Location
Chr.18: 36350895 - 36352308 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062163.1 XP_017917652.1 2 - 3 693 91

View Full Product Details