Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
102181178
Gene Name
chitinase 3 like 2
Gene Aliases
-
Chromosome Location
Chr.3: 88414258 - 88429954 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677907.3 XP_005677964.1 3 - 4 344 69

View Full Product Details