Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB8
Entrez Gene ID
102182717
Gene Name
heat shock protein family B (small) member 8
Gene Aliases
-
Chromosome Location
Chr.17: 15082188 - 15095210 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691456.3 XP_005691513.1 1 - 2 842 67
GenBank mRNA JO634084.2 - 1 - 2 965 67

View Full Product Details