Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRR19
Entrez Gene ID
102172652
Gene Name
proline rich 19
Gene Aliases
-
Chromosome Location
Chr.18: 51865060 - 51868015 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692564.3 XP_005692621.3 2 - 3 732 110

View Full Product Details