Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOC2B
Entrez Gene ID
102174060
Gene Name
double C2 domain beta
Gene Aliases
-
Chromosome Location
Chr.19: 22271682 - 22297743 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064376.1 XP_017919865.1 6 - 7 1076 64

View Full Product Details