Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXL16
Entrez Gene ID
102174526
Gene Name
F-box and leucine rich repeat protein 16
Gene Aliases
-
Chromosome Location
Chr.25: 533924 - 545442 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041034.1 XP_017896523.1 2 - 3 988 64
GenBank mRNA JO631858.2 - 2 - 3 1026 64

View Full Product Details