Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JHY
Entrez Gene ID
102181698
Gene Name
junctional cadherin complex regulator
Gene Aliases
C15H11orf63
Chromosome Location
Chr.15: 48747206 - 48827933 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689564.3 XP_005689621.2 7 - 8 2186 98

View Full Product Details