Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD3
Entrez Gene ID
102174949
Gene Name
major facilitator superfamily domain containing 3
Gene Aliases
-
Chromosome Location
Chr.14: 81498024 - 81500295 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058774.1 XP_017914263.1 1 - 2 1167 94
XM_018058776.1 XP_017914265.1 1 - 2 1167 94
XM_018058780.1 XP_017914269.1 1 - 2 1167 94

View Full Product Details