Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF6
Entrez Gene ID
102189494
Gene Name
Kruppel like factor 6
Gene Aliases
-
Chromosome Location
Chr.13: 44008662 - 44017430 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057330.1 XP_017912819.1 1 - 2 347 87
GenBank mRNA KA318917.1 - 3 - 4 2578 87
KU041749.1 - 1 - 2 100 87

View Full Product Details