Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LRTM2
Entrez Gene ID
106502149
Gene Name
leucine rich repeats and transmembrane domains 2
Gene Aliases
-
Chromosome Location
Chr.5: 107170905 - 107184796 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048688.1 XP_017904177.1 3 - 4 666 62
GenBank mRNA JO624034.2 - 1 - 2 254 62

View Full Product Details