Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPFF
Entrez Gene ID
102170026
Gene Name
neuropeptide FF-amide peptide precursor
Gene Aliases
-
Chromosome Location
Chr.5: 26257722 - 26261505 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679874.3 XP_005679931.1 2 - 3 3112 56

View Full Product Details