Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H3orf70
Entrez Gene ID
102180539
Gene Name
chromosome 1 C3orf70 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 81664024 - 81788107 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675256.3 XP_005675313.2 1 - 2 426 62
GenBank mRNA JO639279.2 - 1 - 2 498 62
JO639280.2 - 1 - 2 510 62

View Full Product Details