Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXW12
Entrez Gene ID
102173299
Gene Name
F-box and WD repeat domain containing 12
Gene Aliases
-
Chromosome Location
Chr.8: 89819767 - 89855430 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684240.3 XP_005684297.1 7 - 8 1041 65
XM_005684241.3 XP_005684298.1 6 - 7 1000 65
XM_013966237.2 XP_013821691.1 7 - 8 1003 65

View Full Product Details