Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WNT10A
Entrez Gene ID
102177945
Gene Name
Wnt family member 10A
Gene Aliases
-
Chromosome Location
Chr.2: 28944026 - 28957602 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059319.1 XP_017914808.1 2 - 3 820 70
GenBank mRNA KA324023.1 - 2 - 3 1554 70

View Full Product Details