Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102188370
Entrez Gene ID
102188370
Gene Name
Golgi pH regulator
Gene Aliases
GPR89A
Chromosome Location
Chr.3: 98805725 - 98850774 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677751.2 XP_005677808.1 4 - 5 467 109
XM_005677753.3 XP_005677810.1 4 - 5 468 109
XM_018046005.1 XP_017901494.1 2 - 3 168 109
GenBank mRNA JO594747.2 - 4 - 5 426 109
JO646858.2 - 6 - 7 540 109

View Full Product Details