Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF26
Entrez Gene ID
102173217
Gene Name
zinc finger protein 26
Gene Aliases
-
Chromosome Location
Chr.17: 27126935 - 27147246 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061469.1 XP_017916958.1 2 - 3 264 96
XM_018061470.1 XP_017916959.1 3 - 4 633 96

View Full Product Details