Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF84
Entrez Gene ID
108637887
Gene Name
zinc finger protein 84
Gene Aliases
-
Chromosome Location
Chr.17: 27081230 - 27095979 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061461.1 XP_017916950.1 3 - 4 371 78

View Full Product Details